Anti-Theophylline Rabbit Polyclonal Antibody

Lieferant: US Biological
T4500-05A
USBIT4500-05AEA 463 EUR
USBIT4500-05A
Anti-Theophylline Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Theophylline
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR