Anti-THOP1 Mouse Monoclonal Antibody [clone: 9F204]

Lieferant: US Biological
T5008-76C
USBIT5008-76CEA 626 EUR
USBIT5008-76C
Anti-THOP1 Mouse Monoclonal Antibody [clone: 9F204]
Antikörper
Type: Primary
Antigen: THOP1 (thimet oligopeptidase 1)
Clonality: monoclonal
Clone: 9F204
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR