Anti-Threonine Mouse Monoclonal Antibody (Biotin) [clone: 11C156]
Lieferant: US Biological
T5020-05
USBIT5020-05EA 709 EUR
USBIT5020-05
Anti-Threonine Mouse Monoclonal Antibody (Biotin) [clone: 11C156]
Antikörper
Recommended dilution: ELISA: 1:1000-1:5000; Western Blot: 1:500-1:2000
Type: Primary
Antigen: Threonine
Clonality: monoclonal
Clone: 11C156
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen