Anti-Thyroxine Mouse Monoclonal Antibody [clone: 10F530]

Lieferant: US Biological
T5460-01
USBIT5460-01EA 1010 EUR
USBIT5460-01
Anti-Thyroxine Mouse Monoclonal Antibody [clone: 10F530]
Antikörper
Type: Primary
Antigen: Thyroxine
Clonality: monoclonal
Clone: 10F530
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR