Anti-Thyroxine Mouse Monoclonal Antibody [clone: 9L720]

Lieferant: US Biological
T5460-01G
USBIT5460-01GEA 717 EUR
USBIT5460-01G
Anti-Thyroxine Mouse Monoclonal Antibody [clone: 9L720]
Antikörper
Type: Primary
Antigen: Thyroxine
Clonality: monoclonal
Clone: 9L720
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR