Anti-Thyroxin Rabbit Polyclonal Antibody
Lieferant: US Biological
T5460-01J-100
USBIT5460-01J-100EA 939 EUR
USBIT5460-01J-100
Anti-Thyroxin Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Radio Immunoassay: 1:1.000
Type: Primary
Antigen: Thyroxin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen