Anti-Thyroxine Sheep Polyclonal Antibody

Lieferant: US Biological
T5460-01K
USBIT5460-01KEA 479 EUR
USBIT5460-01K
Anti-Thyroxine Sheep Polyclonal Antibody
Antikörper
Recommended dilution: Radio Immunoassay: 1:19

Type: Primary
Antigen: Thyroxine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR