Anti-Thyroxine Mouse Monoclonal Antibody [clone: 9F75]

Lieferant: US Biological
T5460-02A
USBIT5460-02AEA 661 EUR
USBIT5460-02A
Anti-Thyroxine Mouse Monoclonal Antibody [clone: 9F75]
Antikörper
Type: Primary
Antigen: Thyroxine
Clonality: monoclonal
Clone: 9F75
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR