Anti-Thyroxine Binding Globulin Mouse Monoclonal Antibody [clone: 9L722]

Lieferant: US Biological
T5461-01
USBIT5461-01EA 668 EUR
USBIT5461-01
Anti-Thyroxine Binding Globulin Mouse Monoclonal Antibody [clone: 9L722]
Antikörper
Type: Primary
Antigen: Thyroxine Binding Globulin
Clonality: monoclonal
Clone: 9L722
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR