Anti-Thyroxine Binding Globulin Rabbit Polyclonal Antibody

Lieferant: US Biological
T5461-01G
USBIT5461-01GEA 649 EUR
USBIT5461-01G
Anti-Thyroxine Binding Globulin Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-1:100

Type: Primary
Antigen: Thyroxine Binding Globulin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR