Anti-Thyroxine Binding Globulin Goat Polyclonal Antibody
Lieferant: US Biological
T5461-15D
USBIT5461-15DEA 473 EUR
USBIT5461-15D
Anti-Thyroxine Binding Globulin Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Thyroxine Binding Globulin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human