Anti-Thyroxine Binding Globulin Chicken Polyclonal Antibody

Lieferant: US Biological
T5461-16-200
USBIT5461-16-200EA 244 EUR
USBIT5461-16-200
Anti-Thyroxine Binding Globulin Chicken Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Thyroxine Binding Globulin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR