Anti-Thyroxine Binding Globulin Mouse Monoclonal Antibody [clone: 8.F.274]
Lieferant: US Biological
T5461-500
USBIT5461-500EA 729 EUR
USBIT5461-500
Anti-Thyroxine Binding Globulin Mouse Monoclonal Antibody [clone: 8.F.274]
Antikörper
Type: Primary
Antigen: Thyroxine Binding Globulin
Clonality: monoclonal
Clone: 8.F.274
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human