Anti-Tobramycin Mouse Monoclonal Antibody [clone: 9L727]

Lieferant: US Biological
T8000-10A
USBIT8000-10AEA 627 EUR
USBIT8000-10A
Anti-Tobramycin Mouse Monoclonal Antibody [clone: 9L727]
Antikörper
Type: Primary
Antigen: Tobramycin
Clonality: monoclonal
Clone: 9L727
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR