Anti-Tobramycin Sheep Polyclonal Antibody
Lieferant: US Biological
T8000-14
USBIT8000-14EA 1020 EUR
USBIT8000-14
Anti-Tobramycin Sheep Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1 ?g/ml
Type: Primary
Antigen: Tobramycin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity:
Bestellen