Anti-TLR1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
T8050-01A-BIOT
USBIT8050-01A-BIOTEA 884 EUR
USBIT8050-01A-BIOT
Anti-TLR1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: TLR1 (toll-like receptor 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR