Anti-TRAF2 Rabbit Polyclonal Antibody

Lieferant: US Biological
T8171-94
USBIT8171-94EA 647 EUR
USBIT8171-94
Anti-TRAF2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5-1 ?g’ml

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR