Anti-TRAF2 Rabbit Polyclonal Antibody
Lieferant: US Biological
T8171-94L
USBIT8171-94LEA 667 EUR
USBIT8171-94L
Anti-TRAF2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50
Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human