Anti-TRAF2 Rabbit Polyclonal Antibody

Lieferant: US Biological
T8171-95B
USBIT8171-95BEA 391 EUR
USBIT8171-95B
Anti-TRAF2 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunoprecipitation: 1:50; Western Blot: 1:1000

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR