Anti-Transcription Factor Dp-2 Mouse Monoclonal Antibody [clone: 8C337]

Lieferant: US Biological
T8195-82
USBIT8195-82EA 473 EUR
USBIT8195-82
Anti-Transcription Factor Dp-2 Mouse Monoclonal Antibody [clone: 8C337]
Antikörper
Type: Primary
Antigen: Transcription Factor Dp-2
Clonality: monoclonal
Clone: 8C337
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR