Anti-Transforming Growth Factor beta 1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K71]

Lieferant: US Biological
T8250-16F1
USBIT8250-16F1EA 687 EUR
USBIT8250-16F1
Anti-Transforming Growth Factor beta 1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 10K71]
Antikörper
Do not freeze!

Type: Primary
Antigen: Transforming Growth Factor beta 1
Clonality: monoclonal
Clone: 10K71
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR