Anti-TRAPPC4 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
T8267-01B-BIOT
USBIT8267-01B-BIOTEA 884 EUR
USBIT8267-01B-BIOT
Anti-TRAPPC4 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: TRAPPC4 (trafficking protein particle complex 4)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR