Anti-Trypsin Inhibitor, Soybean Rabbit Polyclonal Antibody

Lieferant: US Biological
T8685-05B
USBIT8685-05BEA 584 EUR
USBIT8685-05B
Anti-Trypsin Inhibitor, Soybean Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:50000-1:240000

Type: Primary
Antigen: Trypsin Inhibitor, Soybean
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR