Anti-TNFRSF1B Rat Monoclonal Antibody [clone: HM102]

Lieferant: US Biological
T9161-22V
USBIT9161-22VEA 708 EUR
USBIT9161-22V
Anti-TNFRSF1B Rat Monoclonal Antibody [clone: HM102]
Antikörper
Recommended dilution: Immunochemistry: 1:10; Flow Cytometry: 1:10

Type: Primary
Antigen: TNFRSF1B (tumor necrosis factor receptor superfamily, member 1B)
Clonality: monoclonal
Clone: HM102
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR