Anti-UBQLN1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
U1000-17B-BIOT
USBIU1000-17B-BIOTEA 884 EUR
USBIU1000-17B-BIOT
Anti-UBQLN1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: UBQLN1 (ubiquilin 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR