Anti-UBE2C Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
U1000-86B-BIOT
USBIU1000-86B-BIOTEA 884 EUR
USBIU1000-86B-BIOT
Anti-UBE2C Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR