Anti-UBE2C Goat Polyclonal Antibody
Lieferant: US Biological
U1000-86E
USBIU1000-86EEA 601 EUR
USBIU1000-86E
Anti-UBE2C Goat Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:32000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: UBE2C (ubiquitin-conjugating enzyme E2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human