Anti-UHRF1 Rabbit Polyclonal Antibody
Lieferant: US Biological
U1008-01
USBIU1008-01EA 667 EUR
USBIU1008-01
Anti-UHRF1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1:500
Type: Primary
Antigen: UHRF1 (ubiquitin-like with PHD and ring finger domains 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human