Anti-UHRF1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
U1008-01-BIOT
USBIU1008-01-BIOTEA 884 EUR
USBIU1008-01-BIOT
Anti-UHRF1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: UHRF1 (ubiquitin-like with PHD and ring finger domains 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR