Anti-UBX domain-containing protein 1 Rabbit Polyclonal Antibody

Lieferant: US Biological
U1061
USBIU1061EA 564 EUR
USBIU1061
Anti-UBX domain-containing protein 1 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.1 ?g/ml

Type: Primary
Antigen: UBX domain-containing protein 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR