Anti-UGT1A1 Goat Polyclonal Antibody
Lieferant: US Biological
U1275-27
USBIU1275-27EA 870 EUR
USBIU1275-27
Anti-UGT1A1 Goat Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1 ?g/ml
Type: Primary
Antigen: UGT1A1 (UDP glucuronosyltransferase 1 family, polypeptide A1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human