Anti-FERMT3 Rabbit Polyclonal Antibody

Lieferant: US Biological
U1610-03
USBIU1610-03EA 580 EUR
USBIU1610-03
Anti-FERMT3 Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 0.5 ?g/ml; Immunochemistry: 1:500

Type: Primary
Antigen: FERMT3 (fermitin family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR