Anti-Uridine-5’-Diphosphoglucose Dehydrogenase Rabbit Polyclonal Antibody (Biotin)

Lieferant: US Biological
U2279-03B
USBIU2279-03BEA 626 EUR
USBIU2279-03B
Anti-Uridine-5’-Diphosphoglucose Dehydrogenase Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: Uridine-5’-Diphosphoglucose Dehydrogenase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR