Anti-USP18 Rabbit Monoclonal Antibody [clone: 10g92]

Lieferant: US Biological
U4100-85A
USBIU4100-85AEA 608 EUR
USBIU4100-85A
Anti-USP18 Rabbit Monoclonal Antibody [clone: 10g92]
Antikörper
Recommended dilution: Western Blot: 1:1000; Immunoprecipitation: 1:50

Type: Primary
Antigen: USP18 (ubiquitin specific peptidase 18)
Clonality: monoclonal
Clone: 10g92
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR