Anti-Verotoxin I-B Subunit Mouse Monoclonal Antibody [clone: 13C4]
Lieferant: US Biological
V2121-60D
USBIV2121-60DEA 1040 EUR
USBIV2121-60D
Anti-Verotoxin I-B Subunit Mouse Monoclonal Antibody [clone: 13C4]
Antikörper
Recommended dilution: Western Blot: 1:10
Type: Primary
Antigen: Verotoxin I-B Subunit
Clonality: monoclonal
Clone: 13C4
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen