Anti-VTN Mouse Monoclonal Antibody [clone: BE10]

Lieferant: US Biological
V2200-01L
USBIV2200-01LEA 938 EUR
USBIV2200-01L
Anti-VTN Mouse Monoclonal Antibody [clone: BE10]
Antikörper
Type: Primary
Antigen: VTN (vitronectin)
Clonality: monoclonal
Clone: BE10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR