Anti-Factor VIII Related Antigen Rabbit Polyclonal Antibody

Lieferant: US Biological
V2700-04C
USBIV2700-04CEA 373 EUR
USBIV2700-04C
Anti-Factor VIII Related Antigen Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Factor VIII Related Antigen
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR