Anti-Wolf-Hirschhorn Syndrome Candidate 1-like 1 Mouse Monoclonal Antibody [clone: 8G3]

Lieferant: US Biological
W0484-100
USBIW0484-100EA 511 EUR
USBIW0484-100
Anti-Wolf-Hirschhorn Syndrome Candidate 1-like 1 Mouse Monoclonal Antibody [clone: 8G3]
Antikörper
Recommended dilution: Western Blot: 1:1000-1:3000

Type: Primary
Antigen: Wolf-Hirschhorn Syndrome Candidate 1-like 1
Clonality: Monoclonal
Clone: 8G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human,
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR