Anti-Wolf-Hirschhorn Syndrome Candidate 2 Protein Chicken Polyclonal Antibody

Lieferant: US Biological
W0485-01
USBIW0485-01EA 474 EUR
USBIW0485-01
Anti-Wolf-Hirschhorn Syndrome Candidate 2 Protein Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: Wolf-Hirschhorn Syndrome Candidate 2 Protein
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR