Anti-West Nile Virus, Core Rabbit Polyclonal Antibody

Lieferant: US Biological
W1018-40B
USBIW1018-40BEA 596 EUR
USBIW1018-40B
Anti-West Nile Virus, Core Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: West Nile Virus, Core
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR