Anti-West Nile Virus, Core Rabbit Polyclonal Antibody

Lieferant: US Biological
W1018-40C
USBIW1018-40CEA 568 EUR
USBIW1018-40C
Anti-West Nile Virus, Core Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1 ?g/ml

Type: Primary
Antigen: West Nile Virus, Core
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR