Anti-West Nile Virus, Core Rabbit Polyclonal Antibody

Lieferant: US Biological
W1018-40D
USBIW1018-40DEA 715 EUR
USBIW1018-40D
Anti-West Nile Virus, Core Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1 ?g/ml; Immunofluorescence: 20 ?g/ml; Western Blot: 1 ?g/ml

Type: Primary
Antigen: West Nile Virus, Core
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR