Anti-West Nile Virus, Envelope Protein E Rabbit Polyclonal Antibody
Lieferant: US Biological
W1018-50B
USBIW1018-50BEA 674 EUR
USBIW1018-50B
Anti-West Nile Virus, Envelope Protein E Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1-3?g/ml; Immunochemistry: 1 ?g/ml
Type: Primary
Antigen: West Nile Virus, Envelope Protein E
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen