Anti-West Nile Virus, Envelope Rabbit Polyclonal Antibody

Lieferant: US Biological
W1018-50G
USBIW1018-50GEA 660 EUR
USBIW1018-50G
Anti-West Nile Virus, Envelope Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: West Nile Virus, Envelope
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR