Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 10D33]

Lieferant: US Biological
W1018-50K
USBIW1018-50KEA 513 EUR
USBIW1018-50K
Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 10D33]
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200

Type: Primary
Antigen: West Nile Virus
Clonality: monoclonal
Clone: 10D33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR