Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 10D34]
Lieferant: US Biological
W1018-50M
USBIW1018-50MEA 513 EUR
USBIW1018-50M
Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 10D34]
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200
Type: Primary
Antigen: West Nile Virus
Clonality: monoclonal
Clone: 10D34
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen