Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 10D36]

Lieferant: US Biological
W1018-50P
USBIW1018-50PEA 478 EUR
USBIW1018-50P
Anti-West Nile Virus Mouse Monoclonal Antibody [clone: 10D36]
Antikörper
Recommended dilution: Immunofluorescence: 1:10-1:50; ELISA: 1:20-1:200

Type: Primary
Antigen: West Nile Virus
Clonality: monoclonal
Clone: 10D36
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR