Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody
Lieferant: US Biological
W1018-52C
USBIW1018-52CEA 581 EUR
USBIW1018-52C
Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1 ?g/ml
Type: Primary
Antigen: West Nile Virus, Matrix
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen