Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody

Lieferant: US Biological
W1018-52D
USBIW1018-52DEA 581 EUR
USBIW1018-52D
Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: ELISA: 1 ?g/ml

Type: Primary
Antigen: West Nile Virus, Matrix
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR