Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody
Lieferant: US Biological
W1018-52G
USBIW1018-52GEA 626 EUR
USBIW1018-52G
Anti-West Nile Virus, Matrix Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-2?g/ml; Immunochemistry: 1-2?g/ml
Type: Primary
Antigen: West Nile Virus, Matrix
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen